Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
Description
Includes
Two handles (18 and 24 in. long), one polyester foam pad, and six TechniCloth™ polyester mop covers. Mop with telescoping handle includes six polyester mop covers.
- Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
- Made from autoclavable Celcon™ plastic
- Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Specifications
| Product Type | Replacement Mop Covers |
| Quantity | 150/Cs. |
| For Use With (Application) | For Cleanroom Class 4 to 8 |
| Material | Polyester |