Learn More
Texwipe™ AlphaMop™ Replacement Pad
Description
Specifications
Specifications
| Product Type | Replacement Cleaning Pad |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Quantity | 25 |
| Includes | Fastening Pins |
| For Use With (Equipment) | AlphaMops TX7108 and TX7108A |
| For Use With (Application) | For Ultraclean Production Environments |
| Material | Polyester |