Learn More
Texwipe™ AlphaMop™
Catalog No. 19037712
Change view
Click to view available options
Material:
Polyester
Product Type:
Cleanroom Mop
Pre-wetted Mop Cover
Pre-wetted Refills Mop Cover
Replacement Mop Head, Foam Pad
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
19037712 Polyester Replacement Mop Head, Foam Pad
19034207 Polyester Cleanroom Mop
191304546 Polyester Pre-wetted Refills Mop Cover
18361653 Polyester Pre-wetted Mop Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19037712 Supplier Texwipe™ Supplier No. TX7118
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Flat mopping system for easy and efficient cleaning of floors, walls and ceilings
Specifically designed for the needs of ultraclean production environments. Easily changeable, laundered polyester mop covers fit over a flat, foam-padded head assembly. Lightweight, durable fiberglass handle provides excellent control over swivel head and accesses hard-to-reach areas with ease.
  • Enables rapid change of wiping surfaces for adherence to strict wiping protocol ISO Class 4 cleanroom-laundered Alpha™ polyester mop cover
  • Replaceable foam pad ensures that the polyester mop cover conforms to textured floors and other surfaces
  • Maintains ultralow generation of particles and extractables Resists corrosion, abrasion and commonly used cleaning and disinfecting chemicals such as bleaches, phenols and quaternary ammonium compound
  • AlphaMop polyester replacement mopheads and foam pads available separately (150 heads and six pads per case)
  • AlphaMop mop covers presaturated with 6% IPA Honeycomb™ 10 double-knit polyester mop covers have an abrasive texture (supplied 150 covers per case)
Specifications
Product Type Replacement Mop Head, Foam Pad
Sterility Non-sterile
Quantity 50 Mop Covers, 6 Pads
Includes 50 mop covers and 6 pads
For Use With (Application) Designed for the needs of Ultraclean Production Environments
Material Polyester