missing translation for 'onlineSavingsMsg'
Learn More

Tetrazolium Violet 98.0+%, TCI America™

Catalog No. p-7136121
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Catalog No. Quantity
T01741G 1 g
T017410G 10 g
2 options

Catalog No. T01741G

Supplier: TCI America T01741G

Only null left

Chemical Identifiers

CAS 1719-71-7
Molecular Formula C23H17ClN4
Molecular Weight (g/mol) 384.867
MDL Number MFCD00011875
InChI Key RONADMZTCCPLEF-UHFFFAOYSA-M
Synonym tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at
PubChem CID 74395
ChEBI CHEBI:75193
IUPAC Name 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-]

Specifications

Quantity 1 g
Formula Weight 384.87
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrazolium Violet
RTECSNumber XF8080000
TSCA No