Learn More
Thermo Scientific Chemicals Tetrazolium Violet, 98+%
CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol
Supplier: Thermo Scientific Chemicals A1600806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetrazolium Violet | |
| 247°C to 249°C | |
| MFCD00011875 | |
| tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at | |
| C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-] | |
| 384.867 | |
| CHEBI:75193 | |
| ≥98% |
| 1719-71-7 | |
| C23H17ClN4 | |
| 5 g | |
| RONADMZTCCPLEF-UHFFFAOYSA-M | |
| 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride | |
| 74395 | |
| 384.87 |
Chemical Identifiers
| 1719-71-7 | |
| 384.867 | |
| RONADMZTCCPLEF-UHFFFAOYSA-M | |
| 74395 | |
| 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride |
| C23H17ClN4 | |
| MFCD00011875 | |
| tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at | |
| CHEBI:75193 | |
| C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-] |
Safety and Handling
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 217-008-7
RTECSNumber : XF8080000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only