missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrazolium Violet, 98+%

Catalog No. aaa1600803
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
AAA1600803 1 g
AAA1600806 5 g
AAA1600814 25 g
3 options

Catalog No. AAA1600803

Supplier: Thermo Scientific Chemicals A1600803

Only null left

CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1719-71-7
Molecular Formula C23H17ClN4
Molecular Weight (g/mol) 384.867
MDL Number MFCD00011875
InChI Key RONADMZTCCPLEF-UHFFFAOYSA-M
Synonym tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at
PubChem CID 74395
ChEBI CHEBI:75193
IUPAC Name 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-]

Specifications

Melting Point 247°C to 249°C
Quantity 1 g
Formula Weight 384.87
Percent Purity ≥98%
Chemical Name or Material Tetrazolium Violet
Hazard Category H228-H315-H319-H335
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 217-008-7
RTECSNumber XF8080000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only