missing translation for 'onlineSavingsMsg'
Learn More

Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™

CAS: 288-94-8 | CH2N4 | 70.05 g/mol

$376.25 - $376.25

Specifications

Chemical Name or Material Tetrazole
CAS 288-94-8,75-05-8
Color Cream to Orange
Density 0.8000g/mL
Boiling Point 77°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC432991000
View Documents
Thermo Scientific Chemicals
432991000
100 mL AcroSeal™ Glass Bottle
Each for $376.25
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrazole
Cream to Orange
77°C
AcroSeal™ Glass Bottle
0.8
C1=NNN=N1
70.05
CHEBI:33193
Crystalline Powder
288-94-8,75-05-8
0.8000g/mL
5°C
CH2N4
KJUGUADJHNHALS-UHFFFAOYSA-N
2H-tetrazole
67519
70.05
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only