missing translation for 'onlineSavingsMsg'
Learn More

Tetravinyl tin, 95%

Catalog No. AC321080050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC321080050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC321080050 Supplier Thermo Scientific Chemicals Supplier No. 321080050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1112-56-7 | C8H12Sn | 226.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1112-56-7
Molecular Formula C8H12Sn
Molecular Weight (g/mol) 226.88
MDL Number MFCD00008608
InChI Key MZIYQMVHASXABC-UHFFFAOYSA-N
Synonym tetravinyltin, tetravinylstannane, stannane, tetraethenyl, stannane, tetravinyl, tin tetravinyl, sn ch=ch2 4, tetravinyl tin, tetravinyltin iv, tetraethenylstannane, stannane,tetravinyl
PubChem CID 66189
IUPAC Name tetrakis(ethenyl)stannane
SMILES C=C[Sn](C=C)(C=C)C=C

Specifications

Color Colorless to Yellow
Density 1.2400g/mL
Boiling Point 160°C to 163°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5000 to 1.503
Linear Formula Sn(CH=CH2)4
Quantity 5 mL
Specific Gravity 1.24
Formula Weight 226.88
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetravinyl tin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-193-6

RUO – Research Use Only