missing translation for 'onlineSavingsMsg'
Learn More

Tetratriacontane 99.0+%, TCI America™

Catalog No. T29385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29385G 5 g
T29381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29385G Supplier TCI America Supplier No. T29385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14167-59-0
Molecular Formula C34H70
Molecular Weight (g/mol) 478.93
MDL Number MFCD00009412
InChI Key GWVDBZWVFGFBCN-UHFFFAOYSA-N
Synonym n-tetratriacontane, unii-94ua0738yx, tetratriacontane, analytical standard, tetratriacontane, acmc-1bniv
PubChem CID 26519
IUPAC Name tetratriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 74°C
Color White
Boiling Point 285°C
Quantity 5 g
Formula Weight 478.93
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetratriacontane