missing translation for 'onlineSavingsMsg'
Learn More

Tetratriacontane 95.0+%, TCI America™

Catalog No. T06871G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06871G 1 g
T06875G 5 g
T068725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T06871G Supplier TCI America Supplier No. T06871G

Chemical Identifiers

CAS 14167-59-0
Molecular Formula C34H70
Molecular Weight (g/mol) 478.93
MDL Number MFCD00009412
InChI Key GWVDBZWVFGFBCN-UHFFFAOYSA-N
Synonym n-tetratriacontane, unii-94ua0738yx, tetratriacontane, analytical standard, tetratriacontane, acmc-1bniv
PubChem CID 26519
IUPAC Name tetratriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 73°C
Color White
Boiling Point 285°C
Quantity 1 g
Formula Weight 478.93
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetratriacontane