missing translation for 'onlineSavingsMsg'
Learn More

Tetrathiafulvalene 98.0+%, TCI America™

Catalog No. T09805G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09805G 5 g
T09801G 1 g
T098025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T09805G Supplier TCI America Supplier No. T09805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31366-25-3
Molecular Formula C6H4S4
Molecular Weight (g/mol) 204.338
MDL Number MFCD00005492
InChI Key FHCPAXDKURNIOZ-UHFFFAOYSA-N
Synonym tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithiole
PubChem CID 99451
ChEBI CHEBI:52444
IUPAC Name 2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 120°C
Quantity 5 g
Formula Weight 204.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrathiafulvalene