missing translation for 'onlineSavingsMsg'
Learn More

Tetrathiafulvalene (purified by sublimation) 99.0+%, TCI America™

Catalog No. T33801G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33801G 1 g
T3380200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33801G Supplier TCI America Supplier No. T33801G
Only null left

Chemical Identifiers

CAS 31366-25-3
Molecular Formula C6H4S4
Molecular Weight (g/mol) 204.338
MDL Number MFCD00005492
InChI Key FHCPAXDKURNIOZ-UHFFFAOYSA-N
Synonym tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithiole
PubChem CID 99451
ChEBI CHEBI:52444
IUPAC Name 2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 120°C
Quantity 1 g
Formula Weight 204.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrathiafulvalene (purified by sublimation)