missing translation for 'onlineSavingsMsg'
Learn More

Tetrathiafulvalene, 98.5%

Catalog No. p-3737504
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC179412500 250 mg Glass bottle
AC179410010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC179412500 Supplier Thermo Scientific Chemicals Supplier No. 179412500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31366-25-3 | C6H4S4 | 204.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31366-25-3
Molecular Formula C6H4S4
Molecular Weight (g/mol) 204.34
MDL Number MFCD00005492
InChI Key FHCPAXDKURNIOZ-UHFFFAOYSA-N
Synonym tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithiole
PubChem CID 99451
ChEBI CHEBI:52444
IUPAC Name 2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 117°C to 120°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 9390
Formula Weight 204.34
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetrathiafulvalene
Show More Show Less
EINECSNumber 250-593-7

RUO – Research Use Only