Learn More
Tetrathiafulvalene, 97%
CAS: 31366-25-3 | C6H4S4 | 204.338 g/mol
Supplier: Thermo Scientific Chemicals L19229MD
Description
Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.
Solubility
It is insoluble in water. Soluble in organic solvents.
Notes
Air & Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
| Tetrathiafulvalene | |
| 116°C to 120°C | |
| MFCD00005492 | |
| 1617956 | |
| 14,9242 | |
| It is insoluble in water. Soluble in organic solvents. | |
| C1=CSC(=C2SC=CS2)S1 | |
| 204.338 | |
| CHEBI:52444 | |
| 97% |
| 31366-25-3 | |
| C6H4S4 | |
| 250 mg | |
| Air and light sensitive | |
| tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithiole | |
| FHCPAXDKURNIOZ-UHFFFAOYSA-N | |
| 2-(1,3-dithiol-2-ylidene)-1,3-dithiole | |
| 99451 | |
| 204.35 |
Chemical Identifiers
| 31366-25-3 | |
| 204.338 | |
| FHCPAXDKURNIOZ-UHFFFAOYSA-N | |
| 99451 | |
| 2-(1,3-dithiol-2-ylidene)-1,3-dithiole |
| C6H4S4 | |
| MFCD00005492 | |
| tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithiole | |
| CHEBI:52444 | |
| C1=CSC(=C2SC=CS2)S1 |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 250-593-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only