missing translation for 'onlineSavingsMsg'
Learn More

Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex 98.0+%, TCI America™

Catalog No. T24681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24681G Supplier TCI America Supplier No. T24681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40210-84-2
Molecular Formula C18H8N4S4
Molecular Weight (g/mol) 408.53
MDL Number MFCD03791115
InChI Key OIXMVDHMELKBDX-UHFFFAOYSA-N
Synonym TTF - TCNQ Complex
PubChem CID 9844358
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile;2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N.C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 226°C
Color Black-Purple
Quantity 1 g
UN Number 3439
Formula Weight 408.53
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex