missing translation for 'onlineSavingsMsg'
Learn More

Tetratetracontane 97.0+%, TCI America™

Catalog No. T06291G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06291G 1 g
T0629100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06291G Supplier TCI America Supplier No. T06291G
Only null left

Chemical Identifiers

CAS 7098-22-8
Molecular Formula C44H90
Molecular Weight (g/mol) 619.204
MDL Number MFCD00015268
InChI Key KMXFZRSJMDYPPG-UHFFFAOYSA-N
Synonym n-tetratetracontane, tetratetracontane, analytical standard, acmc-209ogu, tetratetracontane, tetratetracontane, purum gc
PubChem CID 23494
ChEBI CHEBI:84289
IUPAC Name tetratetracontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 86°C
Color White
Quantity 1 g
Formula Weight 619.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetratetracontane