missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetratetracontane 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 619.20 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tetratetracontane |
Safety and Handling
| TSCA | Yes |