missing translation for 'onlineSavingsMsg'
Learn More

Tetrasodium 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate 98.0+%, TCI America™

Catalog No. p-7137446
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
T2844200MG 200 mg
T28441G 1 g
2 options

Catalog No. T2844200MG

Supplier: TCI America T2844200MG

Only null left

Chemical Identifiers

CAS 126824-24-6
Molecular Formula C22H20N2Na4O10
Molecular Weight (g/mol) 564.37
MDL Number MFCD02683087
InChI Key ZWSMLJACYSGFKD-UHFFFAOYSA-J
Synonym bapta-na4, bapta tetrasodium, bapta tetrasodium salt, sodium 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, bapta, tetrasodium salt, bapta tetrasodium salt hydrate, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrasodium salt, tetrasodium 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, 1,2-bis 2-aminophenoxy-ethane-n,n,n, n-tetraacetic acid tetrasodium salt, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid tetrasodium salt hplc
PubChem CID 2734965
IUPAC Name tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 200 mg
Formula Weight 564.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrasodium 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetate
TSCA No
Recommended Storage Freezer