missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropyl Orthosilicate 98.0+%, TCI America™

Catalog No. T117425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T117425ML 25 mL
T1174500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T117425ML Supplier TCI America Supplier No. T117425ML

Chemical Identifiers

CAS 682-01-9
Molecular Formula C12H28O4Si
Molecular Weight (g/mol) 264.437
MDL Number MFCD00009364
InChI Key ZQZCOBSUOFHDEE-UHFFFAOYSA-N
Synonym tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane
PubChem CID 69627
IUPAC Name tetrapropyl silicate
SMILES CCCO[Si](OCCC)(OCCC)OCCC

Specifications

Color Colorless
Boiling Point 225°C
Quantity 25 mL
UN Number 1760
Formula Weight 264.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrapropyl Orthosilicate
EINECSNumber (2)-2048
TSCA Yes