missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropenylsuccinic Anhydride (mixture of branched chain isomers) 85.0+%, TCI America™
$68.97 - $227.41
Chemical Identifiers
| CAS | 26544-38-7 |
|---|---|
| Molecular Formula | C16H26O3 |
| MDL Number | MFCD00005528 |
| Synonym | Dodecenylsuccinic Anhydride, Isododecenylsuccinic Anhydride |
Chemical Identifiers
| 26544-38-7 | |
| C16H26O3 | |
| MFCD00005528 | |
| Dodecenylsuccinic Anhydride, Isododecenylsuccinic Anhydride |
Specifications
| 26544-38-7 | |
| C16H26O3 | |
| 25 g | |
| 266.38 | |
| Liquid |
| Yellow | |
| MFCD00005528 | |
| Dodecenylsuccinic Anhydride, Isododecenylsuccinic Anhydride | |
| ≥85.0% (T) | |
| Tetrapropenylsuccinic Anhydride (mixture of branched chain isomers) |
Safety and Handling
EINECSNumber : (2)-0852
RTECSNumber : WN1300000
TSCA : Yes