missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylsilane 97.0+%, TCI America™

Catalog No. p-7137281
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25541G 1 g
T25545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25541G Supplier TCI America Supplier No. T25541G
Only null left

Chemical Identifiers

CAS 1048-08-4
Molecular Formula C24H20Si
Molecular Weight (g/mol) 336.509
MDL Number MFCD00014069
InChI Key JLAVCPKULITDHO-UHFFFAOYSA-N
Synonym silane, tetraphenyl, tetraphenylsilicon, benzene, 1,1',1,1'-silanetetrayltetrakis, silane, tetraphenyl-,
PubChem CID 66104
IUPAC Name tetraphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 238°C
Color White
Boiling Point 166°C
Quantity 1 g
Formula Weight 336.51
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylsilane