missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylsilane 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 238°C |
| Color | White |
| Boiling Point | 166°C |
| Quantity | 5 g |
| Formula Weight | 336.51 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tetraphenylsilane |
Safety and Handling
| TSCA | Yes |