missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylgermane 95.0+%, TCI America™

Catalog No. T11591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11591G Supplier TCI America Supplier No. T11591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1048-05-1
Molecular Formula C24H20Ge
Molecular Weight (g/mol) 381.054
MDL Number MFCD00002997
InChI Key ILEXMONMGUVLRM-UHFFFAOYSA-N
Synonym tetraphenylgermanium, germane, tetraphenyl, tetraphenylgermane, germanium tetraphenyl
PubChem CID 66103
IUPAC Name tetraphenylgermane
SMILES C1=CC=C(C=C1)[Ge](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 235°C
Color White
Quantity 1 g
Formula Weight 381.05
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylgermane