Learn More
Tetraphenylethylene, 98%
CAS: 632-51-9 | C26H20 | 332.446 g/mol
Supplier: Thermo Scientific Chemicals L0022214
Description
Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Tetraphenylethylene | |
| 222°C to 226°C | |
| C26H20 | |
| 25 g | |
| tetraphenylethylene, tetraphenylethene, 1,1,2,2-tetraphenylethene, ethylene, tetraphenyl, 1,1,2,2-tetraphenylethylene, benzene, 1,1',1,1'-1,2-ethenediylidene tetrakis, tetraphenylethylen, 1,2,2-triphenylvinyl benzene, 1,2,2-tetraphenylethylene, 1,1',1,1'-1,2-ethenediylidene tetrakisbenzene | |
| JLZUZNKTTIRERF-UHFFFAOYSA-N | |
| 1,2,2-triphenylethenylbenzene | |
| 69437 | |
| 98% |
| 632-51-9 | |
| 420°C | |
| MFCD00004764 | |
| 1913548 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4 | |
| 332.446 | |
| 332.45 |
Chemical Identifiers
| 632-51-9 | |
| 332.446 | |
| JLZUZNKTTIRERF-UHFFFAOYSA-N | |
| 69437 | |
| C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4 |
| C26H20 | |
| MFCD00004764 | |
| tetraphenylethylene, tetraphenylethene, 1,1,2,2-tetraphenylethene, ethylene, tetraphenyl, 1,1,2,2-tetraphenylethylene, benzene, 1,1',1,1'-1,2-ethenediylidene tetrakis, tetraphenylethylen, 1,2,2-triphenylvinyl benzene, 1,2,2-tetraphenylethylene, 1,1',1,1'-1,2-ethenediylidene tetrakisbenzene | |
| 1,2,2-triphenylethenylbenzene |
Safety and Handling
EINECSNumber : 211-179-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only