missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylethylene, 98%

Catalog No. AAL0022206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0022206 5 g
AAL0022214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0022206 Supplier Thermo Scientific Chemicals Supplier No. L0022206

CAS: 632-51-9 | C26H20 | 332.446 g/mol

Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 632-51-9
Molecular Formula C26H20
Molecular Weight (g/mol) 332.446
MDL Number MFCD00004764
InChI Key JLZUZNKTTIRERF-UHFFFAOYSA-N
Synonym tetraphenylethylene, tetraphenylethene, 1,1,2,2-tetraphenylethene, ethylene, tetraphenyl, 1,1,2,2-tetraphenylethylene, benzene, 1,1',1,1'-1,2-ethenediylidene tetrakis, tetraphenylethylen, 1,2,2-triphenylvinyl benzene, 1,2,2-tetraphenylethylene, 1,1',1,1'-1,2-ethenediylidene tetrakisbenzene
PubChem CID 69437
IUPAC Name 1,2,2-triphenylethenylbenzene
SMILES C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 222°C to 226°C
Boiling Point 420°C
Quantity 5 g
Beilstein 1913548
Solubility Information Insoluble in water.
Formula Weight 332.45
Percent Purity 98%
Chemical Name or Material Tetraphenylethylene
EINECSNumber 211-179-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only