missing translation for 'onlineSavingsMsg'
Learn More

Tetrapentylammonium bromide, For ion pair chromatography (AT), ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111015709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015709 Supplier Merck Emd Millipore Supplier No. 8799710GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011856

Specifications

Melting Point 100°C to 101°C
CAS 866-97-7
Linear Formula [CH3(CH2)4]4 N(Br)
Quantity 10 g
Formula Weight 378.47
Percent Purity ≥99.0% (AT)
Grade For ion pair chromatography