missing translation for 'onlineSavingsMsg'
Learn More

Tetrapentacontane, Standard for GC Analysis, MilliporeSigma™ Supelco™

Catalog No. 111022792
Change view
Click to view available options
:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022792 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022792 Supplier Merck Emd Millipore Supplier No. 87992100MG
Only null left

Specifications

CAS 5856-66-6
Melting Point 94°C to 97°C
Molecular Formula C54H110
Linear Formula CH3(CH2)52 CH3
MDL Number MFCD00216751
Quantity 100 mg
UN Number NONH for all modes of transport
Formula Weight 759.45
Percent Purity ≥98% (GC)
Grade Analytical Standard
Physical Form Neat
Show More Show Less