missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylurea, 99%

Catalog No. p-7023167
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1629518 50 g
AAA1629530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1629518 Supplier Thermo Scientific Chemicals Supplier No. A1629518

CAS: 632-22-4 | C5H12N2O | 116.164 g/mol

Tetramethylurea is used as a solvent in dyestuff industries, in condensation reaction and intermediates in surfactant. It is utilized for base catalyzed isomerization and alkylation hydrocyanation due to its low permittivity. It reacts with oxalyl chloride to prepare tetramethyl chloroformamidinium chloride, which is used for the conversion of carboxylic acids and dialkyl phosphates to anhydrides and pyrophosphates respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylurea is used as a solvent in dyestuff industries, in condensation reaction and intermediates in surfactant. It is utilized for base catalyzed isomerization and alkylation hydrocyanation due to its low permittivity. It reacts with oxalyl chloride to prepare tetramethyl chloroformamidinium chloride, which is used for the conversion of carboxylic acids and dialkyl phosphates to anhydrides and pyrophosphates respectively.

Solubility
Miscible with water.Miscible with water, petroleum ether and common solvents.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 632-22-4
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.164
MDL Number MFCD00008319
InChI Key AVQQQNCBBIEMEU-UHFFFAOYSA-N
Synonym tetramethylurea, temur, urea, tetramethyl, n,n,n',n'-tetramethylurea, tetramethyluree, tmu, urea, 1,1,3,3-tetramethyl, urea, n,n,n',n'-tetramethyl, tetramethyluree french, tetramethyl-urea
PubChem CID 12437
ChEBI CHEBI:84278
IUPAC Name 1,1,3,3-tetramethylurea
SMILES CN(C)C(=O)N(C)C

Specifications

Melting Point -1°C
Density 0.969
Boiling Point 176°C to 178°C
Flash Point 75°C (167°F)
Odor Mild
Refractive Index 1.4506
Linear Formula (CH3)2NCON(CH3)2
Quantity 50 g
Beilstein 773898
Merck Index 14,9230
Solubility Information Miscible with water;Miscible with water,petroleum ether and common solvents.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material Tetramethylurea
Show More Show Less
Hazard Category H227-H302-H360D
Hazard Statement GHS H Statement
H360-H302-H227
May damage fertility or the unborn child.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P201-P202-P210-P264b-P270-P281-P301+P312-P308+P313-P330-P370+P378q-P501c
EINECSNumber 211-173-9
RTECSNumber YU2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only