missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsuccinonitrile 98.0+%, TCI America™

Catalog No. T015525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T015525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T015525G Supplier TCI America Supplier No. T015525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3333-52-6
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00040683
InChI Key ZVQXQPNJHRNGID-UHFFFAOYSA-N
Synonym tetramethylsuccinonitrile, butanedinitrile, tetramethyl, 2,2,3,3-tetramethylsuccinonitrile, tetramethylsuccinotrile, tetramethyl succinonitrile, tetramethylsuccinodinitrile, tmsn, succinonitrile, tetramethyl, tetramethylbutanedinitrile, tetramethylsukcinonitril
PubChem CID 18745
IUPAC Name tetramethylbutanedinitrile
SMILES CC(C)(C#N)C(C)(C)C#N

Specifications

Melting Point 171°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 136.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylsuccinonitrile
RTECSNumber WN4025000
TSCA No