missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane, For NMR spectroscopy, ACS reagent, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019212
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019212 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019212 Supplier Merck Emd Millipore Supplier No. 8792025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Environmental

Chemical Identifiers

MDL Number MFCD00008274

Specifications

Boiling Point 26°C to 28°C
Melting Point -99°C
Physical Form Neat
CAS 75-76-3
Quantity 25 mL
Linear Formula Si(CH3)4
Flash Point -27°C - Closed Cup
Formula Weight 88.22
Percent Purity ≥99.5% (GC)
Grade ACS Reagent
Ignition Point 842°F
Vapor Pressure 11.66 psi (20°C)
Density 0.648 g/mL (at 25°C)
Show More Show Less