Learn More
Tetramethylsilane, 99.9%
Description
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
Solubility
Miscible with water, ethanol, ether and most organic solvents. Immiscible with cold sulfuric acid.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 26°C to 27°C |
| Melting Point | -100°C |
| Quantity | 100 g |
| Flash Point | −27°C (−16°F) |
| Odor | Mild |
| Refractive Index | 1.359 |
| Density | 0.646 |
| UN Number | UN2749 |
| Beilstein | 1696908 |
| Merck Index | 14,9229 |
| Show More |
Safety and Handling
| Hazard Category | H224 |
| Hazard Statement | GHS H Statement H224 Extremely flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: TETRAMETHYLSILANE |
| EINECSNumber | 200-899-1 |
| RTECSNumber | VV5706000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only