missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylgermanium, 98%

Catalog No. AAA1693106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1693106 5 g
AAA1693114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1693106 Supplier Thermo Scientific Chemicals Supplier No. A1693106

CAS: 865-52-1 | C4H12Ge | 132.77 g/mol

Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 865-52-1
Molecular Formula C4H12Ge
Molecular Weight (g/mol) 132.77
MDL Number MFCD00014843
InChI Key ZRLCXMPFXYVHGS-UHFFFAOYSA-N
Synonym tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference
PubChem CID 70079
IUPAC Name tetramethylgermane
SMILES C[Ge](C)(C)C

Specifications

Melting Point -88°C
Density 0.975
Boiling Point 42°C to 44°C
Flash Point −37°C (−35°F)
Refractive Index 1.389
Quantity 5 g
UN Number UN1993
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 132.73
Percent Purity 98%
Chemical Name or Material Tetramethylgermanium
Show More Show Less
Hazard Category H224-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: I
EINECSNumber 212-745-0
RTECSNumber LY5345000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only