missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylgermane 98.0+%, TCI America™
$109.05 - $325.46
Chemical Identifiers
| CAS | 865-52-1 |
|---|---|
| Molecular Formula | C4H12Ge |
| Molecular Weight (g/mol) | 132.77 |
| MDL Number | MFCD00014843 |
| InChI Key | ZRLCXMPFXYVHGS-UHFFFAOYSA-N |
| Synonym | tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference |
| PubChem CID | 70079 |
| IUPAC Name | tetramethylgermane |
| SMILES | C[Ge](C)(C)C |
Chemical Identifiers
| 865-52-1 | |
| 132.77 | |
| ZRLCXMPFXYVHGS-UHFFFAOYSA-N | |
| 70079 | |
| C[Ge](C)(C)C |
| C4H12Ge | |
| MFCD00014843 | |
| tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference | |
| tetramethylgermane |
Specifications
| 865-52-1 | |
| 43°C | |
| MFCD00014843 | |
| 1993 | |
| ZRLCXMPFXYVHGS-UHFFFAOYSA-N | |
| tetramethylgermane | |
| 70079 | |
| ≥98.0% (GC) | |
| Tetramethylgermane |
| Colorless | |
| C4H12Ge | |
| 1 g | |
| tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference | |
| C[Ge](C)(C)C | |
| 132.77 | |
| 132.77 | |
| Liquid |
Safety and Handling
RTECSNumber : LY5345000
TSCA : No