missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylgermane 98.0+%, TCI America™

Catalog No. T11585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11585G 5 g
T11581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11585G Supplier TCI America Supplier No. T11585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 865-52-1
Molecular Formula C4H12Ge
Molecular Weight (g/mol) 132.77
MDL Number MFCD00014843
InChI Key ZRLCXMPFXYVHGS-UHFFFAOYSA-N
Synonym tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference
PubChem CID 70079
IUPAC Name tetramethylgermane
SMILES C[Ge](C)(C)C

Specifications

Color Colorless
Boiling Point 43°C
Quantity 5 g
UN Number 1993
Formula Weight 132.77
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetramethylgermane
RTECSNumber LY5345000
TSCA No