missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylethylene, 97%

Catalog No. AAA1320106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1320106 5 g
AAA1320114 25 g
AAA1320122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1320106 Supplier Thermo Scientific Chemicals Supplier No. A1320106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 563-79-1 | C6H12 | 84.162 g/mol

Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.

Solubility
Soluble in water (0.071 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.

Chemical Identifiers

CAS 563-79-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00008897
InChI Key WGLLSSPDPJPLOR-UHFFFAOYSA-N
Synonym 2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps
PubChem CID 11250
IUPAC Name 2,3-dimethylbut-2-ene
SMILES CC(=C(C)C)C

Specifications

Melting Point -75°C
Density 0.705
Boiling Point 72°C to 73°C
Flash Point −17°C (1°F)
Odor Mild
Refractive Index 1.412
Quantity 5 g
UN Number UN3295
Beilstein 1361357
Sensitivity Air Sensitive
Solubility Information Soluble in water (0.071g/L)
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material Tetramethylethylene
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 209-263-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only