missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Triacetoxyborohydride 85.0+%, TCI America™

Catalog No. T15535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15535G 5 g
T155325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15535G Supplier TCI America Supplier No. T15535G
Only null left

Chemical Identifiers

CAS 109704-53-2
Molecular Formula C10H22BNO6
Molecular Weight (g/mol) 263.10
MDL Number MFCD00012196
InChI Key LCFZZOGKVOTFPU-UHFFFAOYSA-N
Synonym tetramethylammonium triacetoxyborohydride, me4nhb oac 3, me4n aco 3bh;, me4nb oac 3h;, me4n bh oac 3;, ch3 4nhb oac ;, me4n ch3co2 3bh;, tetramethylammonium triacetoxyhydroborate, tetramethylammonium triacetoxyborohydride;, bis acetyloxy boranuidyl acetate; tetramethylazanium
PubChem CID 10858300
IUPAC Name bis(acetyloxy)boranuidyl acetate; tetramethylazanium
SMILES C[N+](C)(C)C.CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Color White
Quantity 5 g
UN Number 2813
Formula Weight 263.10
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Triacetoxyborohydride