missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium triacetoxyborohydride, 90+%

Catalog No. p-4813685
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC392850100 10 g
AC392850500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392850100 Supplier Thermo Scientific Chemicals Supplier No. 392850100
Only null left

CAS: 109704-53-2 | C10H22BNO6 | 263.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 109704-53-2
Molecular Formula C10H22BNO6
Molecular Weight (g/mol) 263.10
MDL Number MFCD00012196
InChI Key LCFZZOGKVOTFPU-UHFFFAOYSA-N
Synonym tetramethylammonium triacetoxyborohydride, me4nhb oac 3, me4n aco 3bh;, me4nb oac 3h;, me4n bh oac 3;, ch3 4nhb oac ;, me4n ch3co2 3bh;, tetramethylammonium triacetoxyhydroborate, tetramethylammonium triacetoxyborohydride;, bis acetyloxy boranuidyl acetate; tetramethylazanium
PubChem CID 10858300
IUPAC Name tetramethylazanium;triacetyloxyboron(1-)
SMILES C[N+](C)(C)C.CC(=O)O[BH-](OC(C)=O)OC(C)=O

Specifications

Melting Point 95.0°C to 98.0°C
Assay Percent Range 90+%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: .. Other solubilities: soluble in common organic solvents
Formula Weight 263.1
Percent Purity 90+%
Chemical Name or Material Tetramethylammonium triacetoxyborohydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gases.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr

RUO – Research Use Only