missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Tetrafluoroborate 98.0+%, TCI America™
$108.11 - $108.11
Chemical Identifiers
| CAS | 661-36-9 |
|---|---|
| Molecular Formula | C4H12BF4N |
| Molecular Weight (g/mol) | 160.95 |
| MDL Number | MFCD00011745 |
| InChI Key | XWFABLFRYCHILB-UHFFFAOYSA-N |
| Synonym | tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate |
| PubChem CID | 12621 |
| IUPAC Name | tetrafluoroboranuide; tetramethylazanium |
| SMILES | C[N+](C)(C)C.F[B-](F)(F)F |
Chemical Identifiers
| 661-36-9 | |
| 160.95 | |
| XWFABLFRYCHILB-UHFFFAOYSA-N | |
| 12621 | |
| C[N+](C)(C)C.F[B-](F)(F)F |
| C4H12BF4N | |
| MFCD00011745 | |
| tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate | |
| tetrafluoroboranuide; tetramethylazanium |
Specifications
| 661-36-9 | |
| White | |
| MFCD00011745 | |
| 1759 | |
| XWFABLFRYCHILB-UHFFFAOYSA-N | |
| tetrafluoroboranuide; tetramethylazanium | |
| 12621 | |
| ≥98.0% (N) | |
| Tetramethylammonium Tetrafluoroborate |
| 330°C | |
| C4H12BF4N | |
| 25 g | |
| tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate | |
| C[N+](C)(C)C.F[B-](F)(F)F | |
| 160.95 | |
| 160.95 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186&(2)-3044
RTECSNumber : BS8300000
TSCA : No