missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium tetrafluoroborate, 97%

Catalog No. AAA1743606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1743606 5 g
AAA1743614 25 g
AAA1743622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1743606 Supplier Thermo Scientific Chemicals Supplier No. A1743606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 661-36-9 | C4H12BF4N | 160.95 g/mol

It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

Solubility
Solubility in hot water is almost transparent.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.

Chemical Identifiers

CAS 661-36-9
Molecular Formula C4H12BF4N
Molecular Weight (g/mol) 160.95
MDL Number MFCD00011745
InChI Key XWFABLFRYCHILB-UHFFFAOYSA-N
Synonym tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
PubChem CID 12621
IUPAC Name tetramethylazanium;tetrafluoroborate
SMILES C[N+](C)(C)C.F[B-](F)(F)F

Specifications

Melting Point 417°C to 419°C
Quantity 5 g
Beilstein 3715404
Sensitivity Hygroscopic
Formula Weight 160.95
Percent Purity 97%
Chemical Name or Material Tetramethylammonium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-547-1
RTECSNumber BS8300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only