missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Sulfate 98.0+%, TCI America™
$105.89 - $294.56
Chemical Identifiers
| CAS | 14190-16-0 |
|---|---|
| Molecular Formula | C8H24N2O4S |
| Molecular Weight (g/mol) | 244.35 |
| MDL Number | MFCD00012139 |
| InChI Key | KJFVITRRNTVAPC-UHFFFAOYSA-L |
| PubChem CID | 14746849 |
| IUPAC Name | tetramethylazanium;sulfate |
| SMILES | C[N+](C)(C)C.C[N+](C)(C)C.[O-]S(=O)(=O)[O-] |
Chemical Identifiers
| 14190-16-0 | |
| 244.35 | |
| KJFVITRRNTVAPC-UHFFFAOYSA-L | |
| tetramethylazanium;sulfate |
| C8H24N2O4S | |
| MFCD00012139 | |
| 14746849 | |
| C[N+](C)(C)C.C[N+](C)(C)C.[O-]S(=O)(=O)[O-] |
Specifications
| 14190-16-0 | |
| White | |
| MFCD00012139 | |
| KJFVITRRNTVAPC-UHFFFAOYSA-L | |
| tetramethylazanium;sulfate | |
| 14746849 | |
| ≥98.0% (T) | |
| Tetramethylammonium Sulfate |
| 285°C | |
| C8H24N2O4S | |
| 5 g | |
| C[N+](C)(C)C.C[N+](C)(C)C.[O-]S(=O)(=O)[O-] | |
| 244.35 | |
| 244.35 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : No