missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium perchlorate, 98%

Catalog No. AC420561000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420561000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420561000 Supplier Thermo Scientific Chemicals Supplier No. 420561000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2537-36-2 | C4H12ClNO4 | 173.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst

Chemical Identifiers

CAS 2537-36-2
Molecular Formula C4H12ClNO4
Molecular Weight (g/mol) 173.59
MDL Number MFCD00043207
InChI Key ZCWKIFAQRXNZCH-UHFFFAOYSA-M
Synonym tetramethylammonium perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate, n,n,n-trimethylmethanaminium perchlorate, ammonium, tetramethyl-, perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate 1:1, c4h12n.clo4, acmc-20ajj6, tetramethylazanium perchlorate, tetramethylammonium perchlorate 25g, wln: 2k2&2&2 &wgw
PubChem CID 17337
IUPAC Name tetramethylazanium;perchlorate
SMILES C[N+](C)(C)C.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry base)
Packaging Plastic Bottle
Linear Formula (CH3)4NClO4
Quantity 100 g
Formula Weight 173.6
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetramethylammonium perchlorate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-805-5

RUO – Research Use Only