missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Iodide, TCI America™

Catalog No. T013925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T013925G Supplier TCI America Supplier No. T013925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-58-1
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.05
MDL Number MFCD00011629
InChI Key RXMRGBVLCSYIBO-UHFFFAOYSA-M
Synonym tetramethylammonium iodide, artilacer, yodurtan, banikol, tmai, methanaminium, n,n,n-trimethyl-, iodide, ammonium, tetramethyl-, iodide, methanaminium, n,n,n-trimethyl-, iodide 1:1, tetramethylammoniumiodide, tetramethylammoniumjodid
PubChem CID 6381
IUPAC Name tetramethylazanium iodide
SMILES [I-].C[N+](C)(C)C

Specifications

Color White
Quantity 25 g
Formula Weight 201.05
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Iodide
EINECSNumber (2)-0186
RTECSNumber PA1050000
TSCA Yes