missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium iodide, 99%

Catalog No. aaa1281122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1281122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1281122 Supplier Thermo Scientific Chemicals Supplier No. A1281122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75-58-1 | C4H12IN | 201.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-58-1
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.05
MDL Number MFCD00011629
InChI Key RXMRGBVLCSYIBO-UHFFFAOYSA-M
Synonym tetramethylammonium iodide, artilacer, yodurtan, banikol, tmai, methanaminium, n,n,n-trimethyl-, iodide, ammonium, tetramethyl-, iodide, methanaminium, n,n,n-trimethyl-, iodide 1:1, tetramethylammoniumiodide, tetramethylammoniumjodid
PubChem CID 6381
IUPAC Name tetramethylazanium;iodide
SMILES [I-].C[N+](C)(C)C

Specifications

Melting Point >300°C
Odor Odorless
Linear Formula (CH3)4NI
Quantity 100 g
UN Number UN2811
Beilstein 3620030
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9225
Solubility Information Slightly soluble in water; Insoluble in chloroform, ether
Formula Weight 201.05
Percent Purity 99%
Chemical Name or Material Tetramethylammonium iodide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-881-3
RTECSNumber PA1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only