missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide, 5% in Methanol, Reagents

Catalog No. 01333462
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333462 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333462 Supplier Labchem Inc Supplier No. CT0078001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetramethylammonium Hydroxide, 5% in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-59-2 , 67-56-1
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.15
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
IUPAC Name tetramethylazanium hydroxide
SMILES [OH-].C[N+](C)(C)C

Specifications

Color Colorless to Light Yellow
pH 12 to 13
Boiling Point 65°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.807
Percent Purity 5%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Tetramethylammonium Hydroxide, 5% in Methanol