missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium hydroxide, 25% in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Chemical Name or Material | Tetramethylammonium hydroxide |
| Name Note | 25% in Water |
| CAS | 75-59-2, 7732-18-5 |
| Melting Point | -25.0°C |
| Color | Colorless to Yellow |
| Density | 1.0140g/mL |
| Boiling Point | 102.0°C |
| Flash Point | >95°C |
| Packaging | Plastic bottle |
| Molecular Formula | C4H13NO |
| Show More |
RUO – Research Use Only