missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide, 25% w/w in methanol

Catalog No. p-7049898
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3083309 10 g
AA3083322 100 g
AA3083336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3083309 Supplier Thermo Scientific Chemicals Supplier No. 03083309
Only null left

CAS: 75-59-2 | C4H13NO | 91.154 g/mol

Tetramethylammonium hydroxide is used for anisotropic etching of silicon. As a basic solvent, it is used in the development of acidic photoresist in the photolithography process. It acts as phase transfer catalyst and surfactant and is used in the synthesis of ferrofluid in order to inhibit nanoparticle aggregation. It finds application in thermochemolysis, an analytical technique involving both pyrolysis and chemical derivatization of the analyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-59-2
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
MDL Number MFCD00008280
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Specifications

Color Colorless to Yellow
Density 0.865 g/mL
Flash Point 12°C (54°F)
Odor Like alcohol
Linear Formula (CH3)4NOH
Quantity 10 g
UN Number UN2924
Sensitivity Air Sensitive
Merck Index 14,9224
Solubility Information Miscible with water.
Formula Weight 91.15
Concentration or Composition (by Analyte or Components) 25% w/w in methanol
Physical Form Liquid
Chemical Name or Material Tetramethylammonium hydroxide
Show More Show Less
Hazard Category H225-H300+H310-H314-H331-H335-H336-H370-H372
Hazard Statement GHS H Statement
H225-H300-H310-H331-H370-H372-H314
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 200-882-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only