missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide, 10% in Water

Catalog No. p-198367
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420511000 100 g Plastic bottle
AC420515000 500 g Plastic bottle
AC420510010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC420511000 Supplier Thermo Scientific Chemicals Supplier No. 420511000
Only null left

CAS: 75-59-2 | C4H13NO | 91.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 10% in Water
CAS 75-59-2, 7732-18-5
Color Colorless
Density 1.0000g/mL
Boiling Point 100.0°C
Packaging Plastic bottle
Molecular Formula C4H13NO
Linear Formula (CH3)4NOH
Quantity 100 g
MDL Number MFCD00008280
Beilstein 04, 50
Fieser 11,514
Specific Gravity 1
Merck Index 15, 9371
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
Solubility Information Solubility in water: soluble.
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.15
PubChem CID 60966
Formula Weight 91.15
Percent Purity 9.5 to 10.5%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only