missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide (10% in Methanol), TCI America™

Catalog No. p-7136391
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
T067625ML 25 mL
T0676500ML 500 mL
2 options

Catalog No. T067625ML

Supplier: TCI America T067625ML

Only null left

Chemical Identifiers

CAS 75-59-2
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
MDL Number MFCD00008280
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Specifications

Color Yellow
Quantity 25 mL
UN Number 2924
Formula Weight 91.15
Physical Form Liquid
Chemical Name or Material Tetramethylammonium Hydroxide (10% in Methanol)
EINECSNumber (2)-0186
RTECSNumber PA0875000
TSCA Yes