missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide (10% in Methanol) [for Photoresist Research], TCI America™

Catalog No. T120125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T120125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T120125ML Supplier TCI America Supplier No. T120125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-59-2
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
MDL Number MFCD00008280
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Specifications

Color Yellow
Quantity 25 mL
UN Number 2924
Formula Weight 91.15
Physical Form Liquid
Chemical Name or Material Tetramethylammonium Hydroxide (10% in Methanol) [for Photoresist Research]
EINECSNumber (2)-0186
RTECSNumber PA0875000
TSCA Yes