missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hexafluorophosphate, TCI America™

Catalog No. T129625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T129625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T129625G Supplier TCI America Supplier No. T129625G

Chemical Identifiers

CAS 558-32-7
Molecular Formula C4H12F6NP
Molecular Weight (g/mol) 219.111
MDL Number MFCD00012012
InChI Key ZFKULDQWLXAKIM-UHFFFAOYSA-N
Synonym tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
PubChem CID 11209
IUPAC Name tetramethylazanium;hexafluorophosphate
SMILES C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

Specifications

Color White
Quantity 25 g
Formula Weight 219.11
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Hexafluorophosphate
RTECSNumber BS8000000
TSCA No